Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTTGGACTGGGTGCTGGTGGT[A/G]GAGTCAAAGTGGACCAAGTCAAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611848 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR608 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR608 - microRNA 608 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL43 - mitochondrial ribosomal protein L43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308396.1 | Intron | NP_001295325.1 | ||||
NM_032112.2 | Intron | NP_115488.2 | ||||
NM_176792.2 | Intron | NP_789762.1 | ||||
NM_176793.1 | Intron | NP_789763.1 | ||||
NM_176794.1 | Intron | NP_789764.1 | ||||
XM_005270231.2 | Intron | XP_005270288.1 | ||||
XM_006718035.3 | Intron | XP_006718098.1 | ||||
XM_017016790.1 | Intron | XP_016872279.1 |
SEMA4G - semaphorin 4G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203244.1 | Intron | NP_001190173.1 | ||||
NM_017893.3 | Intron | NP_060363.2 | ||||
XM_005270008.2 | Intron | XP_005270065.1 |
Set Membership: |
HapMap |