Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCGTCCCATGACCTACCCTGTG[A/G]GGTGACACCATGATTCCCCACTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 178635 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMS1P21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMS1P21 - BMS1, ribosome biogenesis factor pseudogene 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFTPD - surfactant protein D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003019.4 | Intron | NP_003010.4 | ||||
XM_011540087.1 | Intron | XP_011538389.1 | ||||
XM_011540088.2 | Intron | XP_011538390.1 |