Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGTTAAGGCCCAAAAATGAAA[A/C]GTTTTGCTTCTCCTACTCCTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614440 | ||||||||||||||||||||
Literature Links: |
PSD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSD3 - pleckstrin and Sec7 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015310.3 | 10498 | UTR 3 | NP_056125.3 | |||
NM_206909.2 | 10498 | UTR 3 | NP_996792.1 | |||
XM_011544467.2 | 10498 | Intron | XP_011542769.1 | |||
XM_011544468.2 | 10498 | Intron | XP_011542770.1 | |||
XM_011544469.2 | 10498 | Intron | XP_011542771.1 | |||
XM_011544471.2 | 10498 | Intron | XP_011542773.1 | |||
XM_011544473.2 | 10498 | Intron | XP_011542775.1 | |||
XM_017013258.1 | 10498 | UTR 3 | XP_016868747.1 | |||
XM_017013259.1 | 10498 | UTR 3 | XP_016868748.1 | |||
XM_017013260.1 | 10498 | UTR 3 | XP_016868749.1 | |||
XM_017013261.1 | 10498 | UTR 3 | XP_016868750.1 | |||
XM_017013262.1 | 10498 | Intron | XP_016868751.1 | |||
XM_017013263.1 | 10498 | Intron | XP_016868752.1 | |||
XM_017013264.1 | 10498 | Intron | XP_016868753.1 | |||
XM_017013265.1 | 10498 | UTR 3 | XP_016868754.1 |