Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTCGACCTCGAGGATCCAGAG[A/G]TGGAGACGGTACTACCTCCCAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103830 | ||||||||||||||||||||
Literature Links: |
AKR1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR1A1 - aldo-keto reductase family 1 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202413.1 | 139 | Intron | NP_001189342.1 | |||
NM_001202414.1 | 139 | Intron | NP_001189343.1 | |||
NM_006066.3 | 139 | UTR 5 | NP_006057.1 | |||
NM_153326.2 | 139 | UTR 5 | NP_697021.1 | |||
XM_011540491.2 | 139 | UTR 5 | XP_011538793.1 | |||
XM_011540492.2 | 139 | Intron | XP_011538794.1 |