Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCCTCCTCCTCCGGCCAGTCCC[A/C]GTTCCTCAAGGAGCCAGTCTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607410 | ||||||||||||||||||||
Literature Links: |
DMBX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMBX1 - diencephalon/mesencephalon homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147192.2 | Intron | NP_671725.1 | ||||
NM_172225.1 | Intron | NP_757379.1 | ||||
XM_011540668.2 | Intron | XP_011538970.1 | ||||
XM_017000289.1 | Intron | XP_016855778.1 |