Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTTAATTAGGTCCTCAGTACT[C/G]CATCTCTTGTATGCAAGGTTCGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616476 | ||||||||||||||||||||
Literature Links: |
AGBL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGBL4 - ATP/GTP binding protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323573.1 | 1655 | Intron | NP_001310502.1 | |||
NM_001323574.1 | 1655 | Intron | NP_001310503.1 | |||
NM_001323575.1 | 1655 | Intron | NP_001310504.1 | |||
NM_032785.3 | 1655 | Intron | NP_116174.3 | |||
XM_011542308.2 | 1655 | Intron | XP_011540610.1 | |||
XM_011542310.2 | 1655 | Intron | XP_011540612.1 | |||
XM_017002595.1 | 1655 | Missense Mutation | TGC,TGG | C,W 480 | XP_016858084.1 | |
XM_017002596.1 | 1655 | Intron | XP_016858085.1 | |||
XM_017002597.1 | 1655 | Intron | XP_016858086.1 | |||
XM_017002598.1 | 1655 | Intron | XP_016858087.1 |