Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCGCAGCGGGCAGCTCCTCATC[C/T]TGCTCCACTTGAACGGGGACGTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608661 MIM: 604471 | ||||||||||||||||||||
Literature Links: |
PODN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PODN - podocan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC1A7 - solute carrier family 1 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287595.1 | 1474 | Silent Mutation | CAA,CAG | Q,Q 596 | NP_001274524.1 | |
NM_001287596.1 | 1474 | Intron | NP_001274525.1 | |||
NM_001287597.1 | 1474 | Missense Mutation | AGA,GGA | R,G 472 | NP_001274526.1 | |
NM_006671.5 | 1474 | Silent Mutation | CAA,CAG | Q,Q 537 | NP_006662.3 | |
XM_011542002.2 | 1474 | Silent Mutation | CAA,CAG | Q,Q 314 | XP_011540304.1 |