Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAAAAGGAAGAGAAACCTAAGC[A/G]AGCAAGCATAACCCCCTCACCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf141 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf141 - chromosome 1 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276351.1 | Intron | NP_001263280.1 | ||||
NM_001276352.1 | Intron | NP_001263281.1 | ||||
XM_011541465.2 | Intron | XP_011539767.1 | ||||
XM_011541466.2 | Intron | XP_011539768.1 | ||||
XM_011541467.1 | Intron | XP_011539769.1 | ||||
XM_011541469.1 | Intron | XP_011539771.1 | ||||
XM_011541472.1 | Intron | XP_011539774.1 | ||||
XM_011541473.2 | Intron | XP_011539775.1 | ||||
XM_011541475.1 | Intron | XP_011539777.1 | ||||
XM_017001276.1 | Intron | XP_016856765.1 | ||||
XM_017001277.1 | Intron | XP_016856766.1 |
IL23R - interleukin 23 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144701.2 | Intron | NP_653302.2 | ||||
XM_005270516.2 | Intron | XP_005270573.1 | ||||
XM_011540790.2 | Intron | XP_011539092.1 | ||||
XM_011540791.2 | Intron | XP_011539093.1 |