Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605199 | ||||||||||||||||||||
Literature Links: |
SFPQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SFPQ - splicing factor proline and glutamine rich | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005066.2 | 2203 | Intron | NP_005057.1 | |||
XM_005271112.4 | 2203 | UTR 3 | XP_005271169.1 | |||
XM_005271113.4 | 2203 | UTR 3 | XP_005271170.1 | |||
XM_005271115.4 | 2203 | UTR 3 | XP_005271172.1 | |||
XM_011541950.2 | 2203 | UTR 3 | XP_011540252.1 | |||
XM_017002053.1 | 2203 | UTR 3 | XP_016857542.1 | |||
XM_017002054.1 | 2203 | UTR 3 | XP_016857543.1 |