Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTGGAGCAAGGCGTGCTCTG[C/T]GTGCTGCCGATGCTGTGGGCAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600742 | ||||||||||||||||||||
Literature Links: |
TGFBR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TGFBR3 - transforming growth factor beta receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195683.1 | 3035 | Silent Mutation | NP_001182612.1 | |||
NM_001195684.1 | 3035 | Silent Mutation | NP_001182613.1 | |||
NM_003243.4 | 3035 | Silent Mutation | NP_003234.2 | |||
XM_006710867.2 | 3035 | Silent Mutation | XP_006710930.1 | |||
XM_006710868.2 | 3035 | Silent Mutation | XP_006710931.1 |