Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCATAATTCATATATTTCCTTA[C/T]TATTGGTCAAAACATTTATGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCLM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCLM - glutamate-cysteine ligase modifier subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308253.1 | Intron | NP_001295182.1 | ||||
NM_002061.3 | Intron | NP_002052.1 | ||||
XM_011541261.2 | Intron | XP_011539563.1 | ||||
XM_017001056.1 | Intron | XP_016856545.1 | ||||
XM_017001057.1 | Intron | XP_016856546.1 |