Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCATAGAGGTCTCTGATACTCCC[A/C]TTCATAGTCACTTTGCTGTCTTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603809 | ||||||||||||||||||||
Literature Links: |
THRAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THRAP3 - thyroid hormone receptor associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321471.1 | Intron | NP_001308400.1 | ||||
NM_001321473.1 | Intron | NP_001308402.1 | ||||
NM_005119.3 | Intron | NP_005110.2 | ||||
XM_005271371.4 | Intron | XP_005271428.1 |