Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCGCCTGGCCCCGTGGTGCTTT[C/T]CAAAAAGCCTAGAAACATCAGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MFSD14A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFSD14A - major facilitator superfamily domain containing 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033055.2 | Intron | NP_149044.2 | ||||
XM_005271118.1 | Intron | XP_005271175.1 | ||||
XM_011541957.1 | Intron | XP_011540259.1 | ||||
XM_017002084.1 | Intron | XP_016857573.1 |