Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGTGGCCCCCTGCTTAGTTCTT[A/C]CTAATCAGAACACTTAATTTACAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191350 MIM: 601772 MIM: 609806 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPAGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DPAGT1 - dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001382.3 | 2292 | UTR 3 | NP_001373.2 | |||
XM_005271422.2 | 2292 | UTR 3 | XP_005271479.1 | |||
XM_011542648.1 | 2292 | UTR 3 | XP_011540950.1 | |||
XM_017017293.1 | 2292 | UTR 3 | XP_016872782.1 | |||
XM_017017294.1 | 2292 | UTR 3 | XP_016872783.1 | |||
XM_017017295.1 | 2292 | UTR 3 | XP_016872784.1 |
H2AFX - H2A histone family member X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMBS - hydroxymethylbilane synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |