Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGATAACCTGGATCCATAGAT[C/T]GTTTATATTCATCATACCTGGTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120353 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMP1 - matrix metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145938.1 | 1357 | Missense Mutation | CAA,CGA | Q,R 339 | NP_001139410.1 | |
NM_002421.3 | 1357 | Missense Mutation | CAA,CGA | Q,R 405 | NP_002412.1 |
WTAPP1 - Wilms tumor 1 associated protein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |