Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCTACTTCTGTGAGGGGAGCAG[C/T]GAGGCAGGGAAACCCTAGGTTTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114220 MIM: 614030 | ||||||||||||||||||||
Literature Links: |
CAPN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN1 - calpain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198868.1 | Intron | NP_001185797.1 | ||||
NM_001198869.1 | Intron | NP_001185798.1 | ||||
NM_005186.3 | Intron | NP_005177.2 | ||||
XM_006718698.1 | Intron | XP_006718761.1 | ||||
XM_011545292.1 | Intron | XP_011543594.1 |
SPDYC - speedy/RINGO cell cycle regulator family member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |