Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCCGCCACTGGTCTCCTGGGC[A/G]CCTAGACAGGCAGAAGTCATCCCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176875 MIM: 603761 MIM: 610831 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130987 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100130987 - uncharacterized LOC100130987 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1CA - protein phosphatase 1 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008709.1 | Intron | NP_001008709.1 | ||||
NM_002708.3 | Intron | NP_002699.1 | ||||
NM_206873.1 | Intron | NP_996756.1 |
RAD9A - RAD9 checkpoint clamp component A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243224.1 | Intron | NP_001230153.1 | ||||
NM_004584.2 | Intron | NP_004575.1 | ||||
XM_006718652.3 | Intron | XP_006718715.1 | ||||
XM_017018097.1 | Intron | XP_016873586.1 | ||||
XM_017018098.1 | Intron | XP_016873587.1 |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |