Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGCAAATCAGCATCAGAGAGAG[A/G]GTTCCCTGAAAGTCAAGAAATAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606548 | ||||||||||||||||||||
Literature Links: |
MS4A6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A6A - membrane spanning 4-domains A6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247999.1 | 819 | Silent Mutation | ACC,ACT | T,T 213 | NP_001234928.1 | |
NM_022349.3 | 819 | Silent Mutation | ACC,ACT | T,T 185 | NP_071744.2 | |
NM_152851.2 | 819 | Missense Mutation | CCT,TCT | P,S 151 | NP_690590.1 | |
NM_152852.2 | 819 | Silent Mutation | ACC,ACT | T,T 185 | NP_690591.1 | |
XM_005274177.2 | 819 | Silent Mutation | ACC,ACT | T,T 213 | XP_005274234.1 | |
XM_006718658.2 | 819 | Silent Mutation | ACC,ACT | T,T 213 | XP_006718721.1 | |
XM_006718660.2 | 819 | Silent Mutation | ACC,ACT | T,T 168 | XP_006718723.1 | |
XM_006718661.2 | 819 | Silent Mutation | ACC,ACT | T,T 168 | XP_006718724.1 | |
XM_011545209.1 | 819 | Intron | XP_011543511.1 | |||
XM_017018123.1 | 819 | Silent Mutation | ACC,ACT | T,T 140 | XP_016873612.1 | |
XM_017018124.1 | 819 | Silent Mutation | ACC,ACT | T,T 140 | XP_016873613.1 | |
XM_017018125.1 | 819 | Intron | XP_016873614.1 |