Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCTGTTACCTGTATTACCTTG[A/G]ACAAGTCCCTAGAGCTCTCTGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600045 MIM: 615844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDB1 - damage specific DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001923.4 | 733 | Intron | NP_001914.3 |
TKFC - triokinase and FMN cyclase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015533.3 | 733 | Intron | NP_056348.2 | |||
XM_011544912.2 | 733 | UTR 5 | XP_011543214.1 | |||
XM_017017517.1 | 733 | UTR 5 | XP_016873006.1 | |||
XM_017017518.1 | 733 | UTR 5 | XP_016873007.1 | |||
XM_017017519.1 | 733 | Intron | XP_016873008.1 | |||
XM_017017520.1 | 733 | UTR 5 | XP_016873009.1 | |||
XM_017017521.1 | 733 | Intron | XP_016873010.1 | |||
XM_017017522.1 | 733 | Intron | XP_016873011.1 | |||
XM_017017523.1 | 733 | Intron | XP_016873012.1 | |||
XM_017017524.1 | 733 | Intron | XP_016873013.1 |