Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACCAGTTTATTAATTAATTATG[A/G]TTTTTTTTTTTTCTGAAATGGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RTN3 - reticulon 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265589.1 | Intron | NP_001252518.1 | ||||
NM_001265590.1 | Intron | NP_001252519.1 | ||||
NM_001265591.1 | Intron | NP_001252520.1 | ||||
NM_006054.3 | Intron | NP_006045.1 | ||||
NM_201428.2 | Intron | NP_958831.1 | ||||
NM_201429.2 | Intron | NP_958832.1 | ||||
NM_201430.2 | Intron | NP_958833.1 | ||||
XM_011544730.2 | Intron | XP_011543032.1 | ||||
XM_011544731.2 | Intron | XP_011543033.1 | ||||
XM_017017091.1 | Intron | XP_016872580.1 | ||||
XM_017017092.1 | Intron | XP_016872581.1 |