Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600566 MIM: 605577 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRXN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NRXN2 - neurexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP2 - RAS guanyl releasing protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098670.1 | 2482 | UTR 3 | NP_001092140.1 | |||
NM_001098671.1 | 2482 | UTR 3 | NP_001092141.1 | |||
NM_001318398.1 | 2482 | UTR 3 | NP_001305327.1 | |||
NM_153819.1 | 2482 | UTR 3 | NP_722541.1 | |||
XM_005273707.3 | 2482 | UTR 3 | XP_005273764.3 | |||
XM_011544718.1 | 2482 | UTR 3 | XP_011543020.1 | |||
XM_011544720.1 | 2482 | UTR 3 | XP_011543022.1 | |||
XM_011544721.1 | 2482 | UTR 3 | XP_011543023.1 | |||
XM_011544722.1 | 2482 | UTR 3 | XP_011543024.1 | |||
XM_011544723.2 | 2482 | UTR 3 | XP_011543025.1 | |||
XM_011544725.2 | 2482 | UTR 3 | XP_011543027.1 | |||
XM_017017082.1 | 2482 | UTR 3 | XP_016872571.1 | |||
XM_017017083.1 | 2482 | UTR 3 | XP_016872572.1 | |||
XM_017017084.1 | 2482 | UTR 3 | XP_016872573.1 | |||
XM_017017085.1 | 2482 | UTR 3 | XP_016872574.1 | |||
XM_017017086.1 | 2482 | UTR 3 | XP_016872575.1 |
Set Membership: |
HapMap |