Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCCATGAATTCTTATTCAAGAT[G/T]TTATTGGAATTATTTGGTTATTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C5orf24 - chromosome 5 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135586.1 | Intron | NP_001129058.1 | ||||
NM_001300894.1 | Intron | NP_001287823.1 | ||||
NM_152409.3 | Intron | NP_689622.2 | ||||
XM_005271889.3 | Intron | XP_005271946.1 | ||||
XM_017009049.1 | Intron | XP_016864538.1 | ||||
XM_017009050.1 | Intron | XP_016864539.1 |