Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGGAGGATCCCTTGAGCCCAA[A/G]AGGTGGAGGCTGTGGTGAGCTGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603470 | ||||||||||||||||||||
Literature Links: |
ASS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASS1 - argininosuccinate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000050.4 | Intron | NP_000041.2 | ||||
NM_054012.3 | Intron | NP_446464.1 | ||||
XM_005272200.3 | Intron | XP_005272257.1 | ||||
XM_011518705.2 | Intron | XP_011517007.1 | ||||
XM_017014729.1 | Intron | XP_016870218.1 |