Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGGAAACCGAGGGAGCCTAGA[C/T]CTGTTCCATTTCAAAGGCTCACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606074 MIM: 601619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBGT1 - globoside alpha-1,3-N-acetylgalactosaminyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282629.1 | 1904 | UTR 3 | NP_001269558.1 | |||
NM_001282632.1 | 1904 | UTR 3 | NP_001269561.1 | |||
NM_001288572.1 | 1904 | UTR 3 | NP_001275501.1 | |||
NM_001288573.1 | 1904 | UTR 3 | NP_001275502.1 | |||
NM_021996.5 | 1904 | UTR 3 | NP_068836.2 |
RALGDS - ral guanine nucleotide dissociation stimulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |