Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATGCACATCCCTGAAGAAGCT[C/T]GTGGTTCAGGCCCCGTCTCTCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM163B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM163B - family with sequence similarity 163 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080515.2 | Intron | NP_001073984.1 | ||||
XM_005272203.4 | Intron | XP_005272260.1 | ||||
XM_005272204.4 | Intron | XP_005272261.1 | ||||
XM_006717233.3 | Intron | XP_006717296.1 | ||||
XM_011518912.2 | Intron | XP_011517214.1 | ||||
XM_011518913.2 | Intron | XP_011517215.1 | ||||
XM_017015017.1 | Intron | XP_016870506.1 | ||||
XM_017015018.1 | Intron | XP_016870507.1 |