Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTTCCTGGCTTGCAGACAGCC[A/G]CCTTCTCGCTGTATCCTCACGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE6B - phosphodiesterase 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000283.3 | Intron | NP_000274.2 | ||||
NM_001145291.1 | Intron | NP_001138763.1 | ||||
NM_001145292.1 | Intron | NP_001138764.1 | ||||
XM_011513473.2 | Intron | XP_011511775.1 | ||||
XM_011513474.2 | Intron | XP_011511776.1 | ||||
XM_011513475.2 | Intron | XP_011511777.1 | ||||
XM_011513476.2 | Intron | XP_011511778.1 | ||||
XM_011513477.2 | Intron | XP_011511779.1 | ||||
XM_011513478.2 | Intron | XP_011511780.1 | ||||
XM_017008283.1 | Intron | XP_016863772.1 | ||||
XM_017008284.1 | Intron | XP_016863773.1 | ||||
XM_017008285.1 | Intron | XP_016863774.1 | ||||
XM_017008286.1 | Intron | XP_016863775.1 | ||||
XM_017008287.1 | Intron | XP_016863776.1 | ||||
XM_017008288.1 | Intron | XP_016863777.1 | ||||
XM_017008289.1 | Intron | XP_016863778.1 |