Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAATTGTTTTTAAAGCGAGTTA[C/T]CCATCATTTTAGTTGTTTTCTCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 252800 MIM: 610130 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDUA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IDUA - iduronidase, alpha-L- | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC26A1 - solute carrier family 26 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022042.3 | Intron | NP_071325.2 | ||||
NM_134425.2 | Intron | NP_602297.1 | ||||
NM_213613.3 | Intron | NP_998778.1 |
Set Membership: |
HapMap |