Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTCTTATCCCTTGAGGCCTTGT[C/T]CGCTTGATATTTTCTTTGCCTAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 141251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMOX2 - heme oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127204.1 | Intron | NP_001120676.1 | ||||
NM_001127205.1 | Intron | NP_001120677.1 | ||||
NM_001127206.2 | Intron | NP_001120678.1 | ||||
NM_001286267.1 | Intron | NP_001273196.1 | ||||
NM_001286268.1 | Intron | NP_001273197.1 | ||||
NM_001286269.1 | Intron | NP_001273198.1 | ||||
NM_001286270.1 | Intron | NP_001273199.1 | ||||
NM_001286271.1 | Intron | NP_001273200.1 | ||||
NM_002134.3 | Intron | NP_002125.3 | ||||
XM_011522473.1 | Intron | XP_011520775.1 | ||||
XM_017023196.1 | Intron | XP_016878685.1 | ||||
XM_017023197.1 | Intron | XP_016878686.1 |
NMRAL1 - NmrA-like family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305141.1 | Intron | NP_001292070.1 | ||||
NM_001305142.1 | Intron | NP_001292071.1 | ||||
NM_020677.4 | Intron | NP_065728.1 | ||||
XM_006720905.3 | Intron | XP_006720968.1 | ||||
XM_006720906.2 | Intron | XP_006720969.1 | ||||
XM_011522567.2 | Intron | XP_011520869.1 | ||||
XM_017023485.1 | Intron | XP_016878974.1 | ||||
XM_017023486.1 | Intron | XP_016878975.1 | ||||
XM_017023487.1 | Intron | XP_016878976.1 | ||||
XM_017023488.1 | Intron | XP_016878977.1 | ||||
XM_017023489.1 | Intron | XP_016878978.1 | ||||
XM_017023490.1 | Intron | XP_016878979.1 | ||||
XM_017023491.1 | Intron | XP_016878980.1 | ||||
XM_017023492.1 | Intron | XP_016878981.1 |