Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAAGAGAGCAGAGGCCCAGGGC[A/G]GGGGAGTGAAAGGGCCAATTTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610503 MIM: 141251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDIP1 - cell death-inducing p53 target 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199054.1 | 1854 | UTR 3 | NP_001185983.1 | |||
NM_001199055.1 | 1854 | UTR 3 | NP_001185984.1 | |||
NM_001199056.1 | 1854 | UTR 3 | NP_001185985.1 | |||
NM_013399.2 | 1854 | UTR 3 | NP_037531.2 |
HMOX2 - heme oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127204.1 | 1854 | Intron | NP_001120676.1 | |||
NM_001127205.1 | 1854 | Intron | NP_001120677.1 | |||
NM_001127206.2 | 1854 | Intron | NP_001120678.1 | |||
NM_001286267.1 | 1854 | Intron | NP_001273196.1 | |||
NM_001286268.1 | 1854 | Intron | NP_001273197.1 | |||
NM_001286269.1 | 1854 | Intron | NP_001273198.1 | |||
NM_001286270.1 | 1854 | Intron | NP_001273199.1 | |||
NM_001286271.1 | 1854 | Intron | NP_001273200.1 | |||
NM_002134.3 | 1854 | Intron | NP_002125.3 | |||
XM_011522473.1 | 1854 | Intron | XP_011520775.1 | |||
XM_017023196.1 | 1854 | Intron | XP_016878685.1 | |||
XM_017023197.1 | 1854 | Intron | XP_016878686.1 |