Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAGGCAATTATCCGAATAATTG[C/G]GGGGGGGGGCACTTTCGCCTGAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600576 | ||||||||||||||||||||
Literature Links: |
GATA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATA4 - GATA binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308093.1 | Intron | NP_001295022.1 | ||||
NM_001308094.1 | Intron | NP_001295023.1 | ||||
NM_002052.4 | Intron | NP_002043.2 | ||||
XM_005272385.4 | Intron | XP_005272442.1 | ||||
XM_005272386.1 | Intron | XP_005272443.1 | ||||
XM_006716248.1 | Intron | XP_006716311.1 | ||||
XM_011543817.2 | Intron | XP_011542119.1 | ||||
XM_011543818.2 | Intron | XP_011542120.1 | ||||
XM_017013312.1 | Intron | XP_016868801.1 |