Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116810 MIM: 184420 | ||||||||||||||||||||
Literature Links: |
CTSB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTSB - cathepsin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317237.1 | 2339 | UTR 3 | NP_001304166.1 | |||
NM_001908.4 | 2339 | UTR 3 | NP_001899.1 | |||
NM_147780.3 | 2339 | UTR 3 | NP_680090.1 | |||
NM_147781.3 | 2339 | UTR 3 | NP_680091.1 | |||
NM_147782.3 | 2339 | UTR 3 | NP_680092.1 | |||
NM_147783.3 | 2339 | UTR 3 | NP_680093.1 |
FDFT1 - farnesyl-diphosphate farnesyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |