Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGCCTGGGTGACAGAGCAAGA[C/T]TGTCTCAAAAAAAAAAGGAAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMSAP3 - calmodulin regulated spectrin associated protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080429.2 | Intron | NP_001073898.1 | ||||
NM_020902.1 | Intron | NP_065953.1 | ||||
XM_005272491.3 | Intron | XP_005272548.1 | ||||
XM_011528154.2 | Intron | XP_011526456.1 | ||||
XM_017027036.1 | Intron | XP_016882525.1 |