Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGTCCTGTATGTGCAGTGGTCA[A/C]CTGTGCTGGGGTGCTCGAACCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS4 - ceramide synthase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024552.2 | Intron | NP_078828.2 | ||||
XM_011528290.2 | Intron | XP_011526592.1 | ||||
XM_011528291.2 | Intron | XP_011526593.1 | ||||
XM_011528292.2 | Intron | XP_011526594.1 | ||||
XM_011528293.2 | Intron | XP_011526595.1 | ||||
XM_011528294.2 | Intron | XP_011526596.1 | ||||
XM_011528295.2 | Intron | XP_011526597.1 | ||||
XM_017027303.1 | Intron | XP_016882792.1 | ||||
XM_017027304.1 | Intron | XP_016882793.1 | ||||
XM_017027305.1 | Intron | XP_016882794.1 |