Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGTGTTTCCCTGTTTATCCAT[C/T]CCCTGCAAACTGCAGAGTGGCACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 313440 MIM: 305370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4769 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4769 - microRNA 4769 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006950.3 | 666 | Intron | NP_008881.2 | |||
NM_133499.2 | 666 | Intron | NP_598006.1 |
TIMP1 - TIMP metallopeptidase inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003254.2 | 666 | Silent Mutation | ATC,ATT | I,I 158 | NP_003245.1 | |
XM_017029766.1 | 666 | Intron | XP_016885255.1 |