Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD5 - OTU deubiquitinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136157.1 | Intron | NP_001129629.1 | ||||
NM_001136158.1 | Intron | NP_001129630.1 | ||||
NM_001136159.1 | Intron | NP_001129631.1 | ||||
NM_017602.3 | Intron | NP_060072.1 | ||||
XM_005272622.3 | Intron | XP_005272679.1 | ||||
XM_005278039.3 | Intron | XP_005278096.1 | ||||
XM_005278042.4 | Intron | XP_005278099.1 | ||||
XM_006724537.2 | Intron | XP_006724600.1 | ||||
XM_011543932.1 | Intron | XP_011542234.1 | ||||
XM_011543934.2 | Intron | XP_011542236.1 | ||||
XM_017029628.1 | Intron | XP_016885117.1 | ||||
XM_017029629.1 | Intron | XP_016885118.1 | ||||
XM_017029630.1 | Intron | XP_016885119.1 |