Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCTAGGCTGAGCGGCGGAGGTG[C/G]CCGTGCCGCTGGAAGCGGACAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300947 | ||||||||||||||||||||
Literature Links: |
CCDC120 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC120 - coiled-coil domain containing 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163321.2 | 172 | Intron | NP_001156793.2 | |||
NM_001163322.2 | 172 | Intron | NP_001156794.1 | |||
NM_001163323.2 | 172 | Intron | NP_001156795.1 | |||
NM_001271835.1 | 172 | Intron | NP_001258764.1 | |||
NM_001271836.1 | 172 | UTR 5 | NP_001258765.1 | |||
NM_033626.3 | 172 | Intron | NP_296375.1 |