Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGTGCATAATGTTTTGATGTG[C/T]GTAGAAAAAGCTAGTAGCCCAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRBOX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRBOX4 - KRAB box domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129898.1 | Intron | NP_001123370.1 | ||||
NM_001129899.1 | Intron | NP_001123371.1 | ||||
NM_001129900.1 | Intron | NP_001123372.1 | ||||
NM_017776.2 | Intron | NP_060246.2 |