Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTGCTTACTATGGCATCTGGCA[C/G]AAGGAGGACTCCTGATCAATCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300164 MIM: 314370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INE1 - inactivation escape 1 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA1 - ubiquitin like modifier activating enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003334.3 | 922 | Intron | NP_003325.2 | |||
NM_153280.2 | 922 | Intron | NP_695012.1 | |||
XM_005272649.1 | 922 | Intron | XP_005272706.1 | |||
XM_011543954.2 | 922 | Intron | XP_011542256.1 | |||
XM_017029777.1 | 922 | UTR 5 | XP_016885266.1 | |||
XM_017029778.1 | 922 | Intron | XP_016885267.1 | |||
XM_017029779.1 | 922 | Intron | XP_016885268.1 | |||
XM_017029780.1 | 922 | Intron | XP_016885269.1 | |||
XM_017029781.1 | 922 | Intron | XP_016885270.1 |