Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAGCAATCTGCCTGAGCAGTT[C/T]GACTGTGCCATCAACAGCCGCATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAD3B - ATPase family, AAA domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317238.1 | 1674 | Silent Mutation | TTC,TTT | F,F 413 | NP_001304167.1 | |
NM_031921.5 | 1674 | Silent Mutation | TTC,TTT | F,F 459 | NP_114127.3 | |
XM_005244806.3 | 1674 | Missense Mutation | TTC,TTT | F,F 459 | XP_005244863.1 | |
XM_011542241.2 | 1674 | UTR 3 | XP_011540543.1 | |||
XM_011542243.2 | 1674 | Missense Mutation | TTC,TTT | F,F 291 | XP_011540545.1 | |
XM_011542244.1 | 1674 | Missense Mutation | TTC,TTT | F,F 507 | XP_011540546.1 |