Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGCTGACAGCCTTTGTAAACAC[A/G]TTGAATGGCAAAGATGGAACCGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMD4 - proteasome 26S subunit, non-ATPase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002810.2 | Intron | NP_002801.1 | ||||
XM_005245354.2 | Intron | XP_005245411.1 | ||||
XM_006711464.3 | Intron | XP_006711527.1 | ||||
XM_017001856.1 | Intron | XP_016857345.1 |