Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCCCTGAAGATGCCTACCCAT[C/G]CCGTGTGTGTGGTGGGAGTCGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607934 | ||||||||||||||||||||
Literature Links: |
LOC400743 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC400743 - uncharacterized LOC400743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PADI1 - peptidyl arginine deiminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013358.2 | 144 | Missense Mutation | CCC,GCC | P,A 17 | NP_037490.2 | |
XM_011541307.1 | 144 | Missense Mutation | CCC,GCC | P,A 17 | XP_011539609.1 | |
XM_017001101.1 | 144 | Intron | XP_016856590.1 | |||
XM_017001102.1 | 144 | Intron | XP_016856591.1 | |||
XM_017001103.1 | 144 | Missense Mutation | CCC,GCC | P,A 17 | XP_016856592.1 |