Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGCCCGTCCATCCTCAACAGCG[A/G]CCTGGCCAATTTAGGGGCCGAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180480 MIM: 612636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPE - ribulose-5-phosphate-3-epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278282.1 | 141 | UTR 5 | NP_001265211.1 | |||
NM_001278283.1 | 141 | UTR 5 | NP_001265212.1 | |||
NM_001278285.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001265214.1 | |
NM_001278286.1 | 141 | UTR 5 | NP_001265215.1 | |||
NM_001278288.1 | 141 | UTR 5 | NP_001265217.1 | |||
NM_001278289.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001265218.1 | |
NM_001318926.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001305855.1 | |
NM_001318927.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001305856.1 | |
NM_001318928.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001305857.1 | |
NM_001318929.1 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_001305858.1 | |
NM_001318930.1 | 141 | UTR 5 | NP_001305859.1 | |||
NM_001318931.1 | 141 | UTR 5 | NP_001305860.1 | |||
NM_006916.2 | 141 | UTR 5 | NP_008847.1 | |||
NM_199229.2 | 141 | Missense Mutation | GAC,GGC | D,G 15 | NP_954699.1 | |
XM_006712677.3 | 141 | UTR 5 | XP_006712740.1 |
UNC80 - unc-80 homolog, NALCN activator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |