Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCTTCTGATGATGAGTTAATTG[A/G]CCCTCCTTTACCCCCTAAAATGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR70 - WD repeat domain 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018034.2 | 432 | Missense Mutation | GAC,GGC | D,G 119 | NP_060504.1 | |
XM_005248318.3 | 432 | Missense Mutation | GAC,GGC | D,G 118 | XP_005248375.1 | |
XM_011514061.2 | 432 | Intron | XP_011512363.1 | |||
XM_017009604.1 | 432 | Intron | XP_016865093.1 | |||
XM_017009605.1 | 432 | Intron | XP_016865094.1 |