Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGAATTTGCTGCCTGGTGGTG[A/G]TGTTTTTTGTTTTGTTTTGTTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606582 MIM: 612266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLL1 - delta like canonical Notch ligand 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM120B - family with sequence similarity 120B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286379.1 | Intron | NP_001273308.1 | ||||
NM_001286380.1 | Intron | NP_001273309.1 | ||||
NM_001286381.1 | Intron | NP_001273310.1 | ||||
NM_032448.2 | Intron | NP_115824.1 | ||||
XM_011536185.2 | Intron | XP_011534487.1 | ||||
XM_017011357.1 | Intron | XP_016866846.1 | ||||
XM_017011358.1 | Intron | XP_016866847.1 | ||||
XM_017011359.1 | Intron | XP_016866848.1 | ||||
XM_017011360.1 | Intron | XP_016866849.1 |