Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGTGGCTCTGCTTCTAGAAG[C/T]GTAAGGTAACACTGGCATTCCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609685 MIM: 603339 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA7L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDCA7L - cell division cycle associated 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127370.2 | 2557 | UTR 3 | NP_001120842.1 | |||
NM_001127371.2 | 2557 | UTR 3 | NP_001120843.1 | |||
NM_018719.4 | 2557 | UTR 3 | NP_061189.2 |
DNAH11 - dynein axonemal heavy chain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277115.1 | 2557 | Missense Mutation | GCG,GTG | A,V 4516 | NP_001264044.1 |