Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGTTAAGAGAACAGTTCATTA[A/G]AAGTTTCTGTTTAAAAAGAAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612481 MIM: 274180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP12 - poly(ADP-ribose) polymerase family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022750.3 | 3066 | UTR 3 | NP_073587.1 | |||
XM_005250038.3 | 3066 | Intron | XP_005250095.1 | |||
XM_005250039.4 | 3066 | Intron | XP_005250096.1 | |||
XM_005250040.4 | 3066 | Intron | XP_005250097.1 | |||
XM_011516491.2 | 3066 | UTR 3 | XP_011514793.1 |
TBXAS1 - thromboxane A synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |