Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGTGGGACACAGCTGGCCAGGA[A/G]CGCTTCCGCACCATCACCCAAAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB19 - RAB19, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008749.2 | 625 | Silent Mutation | GAA,GAG | E,E 77 | NP_001008749.2 | |
XM_005249996.3 | 625 | Silent Mutation | GAA,GAG | E,E 77 | XP_005250053.1 | |
XM_005249997.3 | 625 | Silent Mutation | GAA,GAG | E,E 65 | XP_005250054.1 | |
XM_005249998.3 | 625 | Silent Mutation | GAA,GAG | E,E 63 | XP_005250055.1 | |
XM_017012207.1 | 625 | Silent Mutation | GAA,GAG | E,E 77 | XP_016867696.1 |
SLC37A3 - solute carrier family 37 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |