Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCACCATGGGGGCTCAAAAAT[A/G]CATTATAATTATAACCATGGTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIP1 - huntingtin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243198.2 | Intron | NP_001230127.1 | ||||
NM_005338.6 | Intron | NP_005329.3 | ||||
XM_005250304.2 | Intron | XP_005250361.1 | ||||
XM_005250305.2 | Intron | XP_005250362.1 | ||||
XM_011516116.2 | Intron | XP_011514418.1 | ||||
XM_017012099.1 | Intron | XP_016867588.1 |