Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACATTTTCAGAGACTGTAACA[A/G]GATCGCATTTTACAGGTAAGGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GANC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GANC - glucosidase alpha, neutral C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301409.1 | 391 | Missense Mutation | AAG,AGG | K,R 26 | NP_001288338.1 | |
NM_001301410.1 | 391 | Missense Mutation | AAG,AGG | K,R 26 | NP_001288339.1 | |
NM_198141.2 | 391 | Missense Mutation | AAG,AGG | K,R 26 | NP_937784.2 |
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |