Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTCGACACTGACGGGCTAGTA[C/G]TGTGCGCCCGCGGGCTCGGCGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 133520 | ||||||||||||||||||||
Literature Links: |
ERCC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERCC4 - ERCC excision repair 4, endonuclease catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005236.2 | 75 | Missense Mutation | CTG,GTG | L,V 34 | NP_005227.1 | |
XM_011522424.2 | 75 | Missense Mutation | CTG,GTG | L,V 34 | XP_011520726.1 | |
XM_011522427.1 | 75 | Intron | XP_011520729.1 | |||
XM_017023043.1 | 75 | UTR 5 | XP_016878532.1 |